Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PCM1 SIRNA Assays ›
Gene Symbol:
PCM1
Gene Name:
pericentriolar material 1
Gene Aliases:
PTC4, RET/PCM-1
Chromosome Location:
Chr.8: 17922763 - 18029948 on Build GRCh38
UniGene ID:
Hs.491148
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s10128
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PCM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001315507.1
NP_001302436.1
4
660
NM_001315508.1
NP_001302437.1
4
660
NM_006197.3
NP_006188.3
4
660
XM_006716336.3
XP_006716399.1
4
733
XM_006716340.3
XP_006716403.1
4
733
XM_006716349.3
XP_006716412.1
4
733
XM_011544528.2
XP_011542830.1
4
733
XM_011544529.2
XP_011542831.1
6
1039
XM_011544530.2
XP_011542832.1
5
855
XM_011544532.2
XP_011542834.1
4
733
XM_011544533.2
XP_011542835.1
4
733
XM_011544534.2
XP_011542836.1
4
733
XM_011544535.2
XP_011542837.1
4
733
XM_011544540.2
XP_011542842.1
5
853
XM_017013472.1
XP_016868961.1
6
1038
XM_017013473.1
XP_016868962.1
6
1037
XM_017013474.1
XP_016868963.1
4
733
XM_017013475.1
XP_016868964.1
6
869
XM_017013476.1
XP_016868965.1
3
453
XM_017013477.1
XP_016868966.1
6
1037
XM_017013478.1
XP_016868967.1
6
1036
XM_017013479.1
XP_016868968.1
6
1037
XM_017013480.1
XP_016868969.1
4
733
XM_017013481.1
XP_016868970.1
6
1036
XM_017013482.1
XP_016868971.1
4
754
XM_017013483.1
XP_016868972.1
6
1035
XM_017013484.1
XP_016868973.1
3
461
XM_017013485.1
XP_016868974.1
6
1036
XM_017013486.1
XP_016868975.1
4
733
XM_017013487.1
XP_016868976.1
4
733
XM_017013488.1
XP_016868977.1
4
733
XM_017013489.1
XP_016868978.1
6
1036
XM_017013490.1
XP_016868979.1
6
1036
XM_017013491.1
XP_016868980.1
4
733
XM_017013492.1
XP_016868981.1
4
733
XM_017013493.1
XP_016868982.1
6
1036
XM_017013494.1
XP_016868983.1
4
733
XM_017013495.1
XP_016868984.1
6
873
XM_017013496.1
XP_016868985.1
3
456
XM_017013497.1
XP_016868986.1
4
733
XM_017013498.1
XP_016868987.1
4
733
XM_017013499.1
XP_016868988.1
6
873
XM_017013500.1
XP_016868989.1
4
733
XM_017013501.1
XP_016868990.1
6
1035
XM_017013502.1
XP_016868991.1
3
456
XM_017013503.1
XP_016868992.1
4
733
XM_017013504.1
XP_016868993.1
4
754
XM_017013505.1
XP_016868994.1
6
873
XM_017013506.1
XP_016868995.1
4
733
XM_017013507.1
XP_016868996.1
6
1037
XM_017013508.1
XP_016868997.1
5
677
GenBank mRNA
AK307583.1
-
4
527
AX785390.1
-
2
238
BC000453.2
-
4
518
BC027477.1
-
3
439
BC065022.1
-
4
511
BC133052.1
-
3
313
8381608
BC140946.1
-
4
489
9056758
L27841.1
-
4
647
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00196390_m1
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
neuron migration
interkinetic nuclear migration
cytoplasmic microtubule organization
microtubule anchoring
microtubule anchoring at centrosome
nonmotile primary cilium assembly
intraciliary transport involved in cilium morphogenesis
cilium assembly
negative regulation of neurogenesis
centrosome organization
protein localization to centrosome
positive regulation of intracellular protein transport
neuronal stem cell population maintenance
protein binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0