Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Shkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138676.2 | NP_619617.2 | 8 | 692 | ||
GenBank mRNA | AF246218.1 | - | 8 | 698 | ||
AK047434.1 | - | 6 | 823 | |||
AK154779.1 | - | 8 | 681 | |||
AK160641.1 | - | 7 | 764 | |||
BC061477.1 | - | 8 | 645 | 6336438 |