Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138677.2 | NP_619618.1 | 7 | 1345 | ||
GenBank mRNA | AB042828.1 | - | 7 | 1334 | ||
AK030943.1 | - | 7 | 1346 | |||
AK037889.1 | - | 7 | 908 | |||
AK082920.1 | - | 7 | 1298 | |||
AK129090.1 | - | 7 | 1312 | |||
AK135969.1 | - | 2 | 209 | |||
AK151668.1 | - | 7 | 1320 | |||
AK152724.1 | - | 7 | 1321 | |||
AK152979.1 | - | 7 | 1317 | |||
BC023237.1 | - | 7 | 1293 | 2654113 |