Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luc7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170848.1 | NP_001164319.1 | 4 | 708 | ||
NM_001170849.1 | NP_001164320.1 | 4 | 708 | |||
NM_138680.2 | NP_619621.2 | 4 | 708 | |||
GenBank mRNA | AF318301.1 | - | 4 | 633 | ||
AK046307.1 | - | 5 | 905 | |||
AK134191.1 | - | 4 | 622 | |||
AK140271.1 | - | 7 | 2837 | |||
AK147889.1 | - | 7 | 879 | |||
AK153527.1 | - | 8 | 965 | |||
BC020058.1 | - | 5 | 433 | 4009362 | ||
BC040793.1 | - | 3 | 200 | |||
BC054449.1 | - | 3 | 651 | |||
BC056354.1 | - | 4 | 706 | 6811460 | ||
BC056383.1 | - | 4 | 687 | 6853955 | ||
BC056970.1 | - | 4 | 706 | 6811460 |