Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luc7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170848.1 | NP_001164319.1 | 8 | 1230 | ||
NM_001170849.1 | NP_001164320.1 | 8 | 1230 | |||
NM_138680.2 | NP_619621.2 | 9 | 1260 | |||
GenBank mRNA | AF318301.1 | - | 9 | 1185 | ||
AK046307.1 | - | 9 | 1428 | |||
AK078853.1 | - | 1 | 104 | |||
AK134191.1 | - | 8 | 1144 | |||
AK138070.1 | - | 1 | 30 | |||
AK147889.1 | - | 11 | 1401 | |||
AK153527.1 | - | 12 | 1487 | |||
AK188016.1 | - | 2 | 101 | |||
BC002314.1 | - | 4 | 195 | |||
BC020058.1 | - | 10 | 985 | 4009362 | ||
BC040793.1 | - | 8 | 752 | |||
BC054449.1 | - | 8 | 1203 | |||
BC056354.1 | - | 9 | 1258 | 6811460 | ||
BC056383.1 | - | 8 | 1209 | 6853955 | ||
BC056970.1 | - | 9 | 1258 | 6811460 |