Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166642.1 | NP_001160114.1 | 15 | 1315 | ||
NM_001166643.1 | NP_001160115.1 | 14 | 1229 | |||
NM_138681.4 | NP_619622.3 | 15 | 1315 | |||
GenBank mRNA | AK032423.1 | - | 15 | 1266 | ||
AK034117.1 | - | 15 | 1295 | |||
AK157222.1 | - | 15 | 1265 | |||
AK160127.1 | - | 14 | 1208 | |||
AK171964.1 | - | 14 | 1182 | |||
BC028339.1 | - | 14 | 1203 | |||
BC038173.1 | - | 13 | 1172 | |||
BC050233.1 | - | 14 | 1208 | |||
BC060690.1 | - | 14 | 1192 | |||
BC080775.1 | - | 13 | 1176 | |||
BC086693.1 | - | 14 | 1196 | |||
BC118965.2 | - | 14 | 1229 | 40097104 | ||
BC118966.1 | - | 14 | 1211 | 40097105 |