Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCSK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000439.4 | NP_000430.3 | 10 | 1546 | ||
NM_001177875.1 | NP_001171346.1 | 10 | 1204 | |||
GenBank mRNA | AK303888.1 | - | 10 | 1204 | ||
AK308575.1 | - | 4 | 452 | |||
BC130295.1 | - | 10 | 1428 | 40146326 | ||
BC136486.1 | - | 10 | 1428 | 9020475 | ||
M90753.1 | - | 10 | 1516 | |||
X64810.1 | - | 10 | 1496 |