Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fchsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146010.1 | NP_001139482.1 | 15, 16 | 1772 | ||
NM_199012.2 | NP_950177.2 | 16, 17 | 1844 | |||
GenBank mRNA | AK122368.1 | - | 16, 17 | 1736 | ||
AK140322.1 | - | 15, 16 | 1751 | |||
AK141949.1 | - | 15, 16 | 1772 | |||
BC034086.1 | - | 3, 4 | 319 | |||
BC039788.1 | - | 2, 3 | 246 | |||
BC060647.1 | - | 16, 17 | 1806 | 6854041 |