Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fchsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146010.1 | NP_001139482.1 | 12 | 1372 | ||
NM_199012.2 | NP_950177.2 | 13 | 1444 | |||
GenBank mRNA | AK122368.1 | - | 13 | 1336 | ||
AK140322.1 | - | 12 | 1351 | |||
AK141949.1 | - | 12 | 1372 | |||
AK162696.1 | - | 1 | 2773 | |||
BC060647.1 | - | 13 | 1406 | 6854041 |