Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172453.3 | NP_766041.1 | 7 | 1885 | ||
GenBank mRNA | AK049353.1 | - | 7 | 1887 | ||
AK084551.1 | - | 1 | 1085 | |||
AK154072.1 | - | 7 | 1931 | |||
AY100322.1 | - | 8 | 1598 | |||
AY498715.1 | - | 7 | 1885 | |||
BC024481.1 | - | 2 | 107 | |||
BC046611.1 | - | 8 | 1523 | 6396273 | ||
BC060108.1 | - | 5 | 2096 |