Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mterf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042670.1 | NP_001036135.2 | 2 | 591 | ||
GenBank mRNA | AK016498.1 | - | 2 | 592 | ||
BC141337.1 | - | 2 | 562 | 9055949 |
Gene Symbol: Mterf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013023.2 | NP_001013041.2 | 2 | 598 | ||
NM_172135.2 | NP_742147.1 | 2 | 579 | |||
GenBank mRNA | AK049308.1 | - | 2 | 599 | ||
AK158221.1 | - | 2 | 1629 | |||
AY726770.2 | - | 2 | 591 | |||
BC038058.1 | - | 2 | 571 | 4207737 | ||
BC051251.1 | - | 2 | 592 | 6490147 | ||
BC082778.1 | - | 2 | 573 | 3994365 | ||
BC094442.1 | - | 2 | 582 | 3587753 |