Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Adamts18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172466.2 | NP_766054.2 | 13 | 2361 | ||
GenBank mRNA | AK036137.1 | - | 10 | 1440 | ||
AK053851.1 | - | 12 | 2473 | |||
AK140876.1 | - | 13 | 2137 | |||
BC094674.1 | - | 13 | 2016 | 30548647 | ||
BC144816.1 | - | 11 | 1619 | |||
BC144817.1 | - | 11 | 1622 |