Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbmx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173376.3 | NP_775552.1 | 4 | 311 | ||
GenBank mRNA | AK076115.1 | - | 4 | 312 | ||
AK160232.1 | - | 4 | 239 | |||
AK168018.1 | - | 4 | 226 | |||
BC026976.1 | - | 4 | 236 | 5361950 |