Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhtkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081131.2 | NP_001074600.1 | 13 | 2381 | ||
GenBank mRNA | AK049272.1 | - | 2372 | |||
AK050057.1 | - | 690 | ||||
BC117994.1 | - | 2332 | 40097991 |