Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166437.1 | NP_001159909.1 | 7 | 1106 | ||
NM_172478.3 | NP_766066.2 | 6 | 1313 | |||
GenBank mRNA | AK040932.1 | - | 6 | 1056 | ||
AK078145.1 | - | 6 | 1314 | |||
AK166256.1 | - | 6 | 1052 | |||
BC060178.1 | - | 5 | 1236 | |||
BC113777.1 | - | 7 | 1054 | |||
BC128565.1 | - | 7 | 1056 | |||
BC128566.1 | - | 6 | 1263 | 40094900 |