Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145952.1 | NP_001139424.1 | 7 | 879 | ||
NM_001145954.1 | NP_001139426.1 | 3 | 293 | |||
NM_178665.5 | NP_848780.3 | 8 | 1086 | |||
GenBank mRNA | AK029335.1 | - | 8 | 878 | ||
AK029567.1 | - | 3 | 294 | |||
AK040643.1 | - | 6 | 640 | |||
AK045288.1 | - | 8 | 947 | |||
AK051345.1 | - | 8 | 958 | |||
AK054550.1 | - | 7 | 882 | |||
AK076989.1 | - | 3 | 284 | |||
AK085626.1 | - | 8 | 1078 | |||
AK086307.1 | - | 8 | 897 | |||
AK168431.1 | - | 8 | 993 | |||
AK169780.1 | - | 8 | 898 | |||
BC085321.1 | - | 7 | 825 | 30743832 |