Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltscr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100452.1 | NP_001093922.1 | 2 | 499 | ||
GenBank mRNA | AK029927.1 | - | 3 | 143 | ||
BC151032.1 | - | 2 | 499 | 9087945 | ||
BC151054.1 | - | 2 | 499 | 9087967 |