Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltscr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100452.1 | NP_001093922.1 | 4 | 1108 | ||
GenBank mRNA | AB093219.1 | - | 1 | 261 | ||
AK029927.1 | - | 5 | 752 | |||
BC151032.1 | - | 4 | 1108 | 9087945 | ||
BC151054.1 | - | 4 | 1108 | 9087967 |