Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpcat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145376.5 | NP_663351.3 | 10, 11 | 1027 | ||
GenBank mRNA | AB244717.2 | - | 10, 11 | 1027 | ||
AK046079.1 | - | 8, 9 | 822 | |||
AK046507.1 | - | 8, 9 | 822 | |||
AK081865.1 | - | 10, 11 | 1185 | |||
AK132379.1 | - | 10, 11 | 1161 | |||
AK155286.1 | - | 10, 11 | 1191 | |||
AK169190.1 | - | 10, 11 | 1160 | |||
AK170723.1 | - | 10, 11 | 1180 | |||
AK171582.1 | - | 10, 11 | 1180 | |||
BC066809.1 | - | 10, 11 | 976 | 30475544 |