Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320613.1 | NP_001307542.1 | 19 | 3450 | ||
NM_004036.4 | NP_004027.2 | 19 | 3447 | |||
XM_006711925.1 | XP_006711988.1 | 20 | 3232 | |||
XM_011532489.1 | XP_011530791.1 | 20 | 3289 | |||
XM_011532490.1 | XP_011530792.1 | 20 | 3286 | |||
XM_011532491.1 | XP_011530793.1 | 19 | 3223 | |||
XM_011532492.2 | XP_011530794.1 | 20 | 3289 | |||
XM_011532493.1 | XP_011530795.1 | 20 | 3151 | |||
XM_011532494.1 | XP_011530796.1 | 20 | 3091 | |||
XM_011532496.1 | XP_011530798.1 | 19 | 2456 | |||
XM_017003186.1 | XP_016858675.1 | 20 | 3229 | |||
XM_017003187.1 | XP_016858676.1 | 19 | 3220 | |||
XM_017003188.1 | XP_016858677.1 | 20 | 3286 | |||
XM_017003189.1 | XP_016858678.1 | 20 | 3148 | |||
XM_017003190.1 | XP_016858679.1 | 19 | 3025 | |||
XM_017003191.1 | XP_016858680.1 | 19 | 2497 | |||
XM_017003192.1 | XP_016858681.1 | 18 | 2333 | |||
XM_017003193.1 | XP_016858682.1 | 18 | 2330 | |||
GenBank mRNA | AB011083.1 | - | 18 | 2283 | ||
AF033861.1 | - | 18 | 3062 | |||
AK027857.1 | - | 18 | 2796 | |||
AK075424.1 | - | 6 | 658 | |||
AK095173.1 | - | 16 | 1856 | |||
AK122926.1 | - | 19 | 3045 | |||
BC126235.1 | - | 18 | 2973 | 8991951 | ||
BC144119.1 | - | 18 | 2976 | 9052639 |