Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfdp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184706.1 | NP_001171635.1 | 12 | 1458 | ||
NM_001184708.1 | NP_001171637.1 | 9 | 1184 | |||
NM_001184709.1 | NP_001171638.1 | 10 | 1082 | |||
NM_001184710.1 | NP_001171639.1 | 9 | 958 | |||
NM_001184711.1 | NP_001171640.1 | 10 | 1065 | |||
NM_178667.4 | NP_848782.2 | 13 | 1459 | |||
GenBank mRNA | AK029055.1 | - | 12 | 1431 | ||
AK033356.1 | - | 10 | 1046 | |||
AK044784.1 | - | 9 | 1186 | |||
AK053881.1 | - | 12 | 1417 | |||
AK079624.1 | - | 13 | 1387 | |||
AK158829.1 | - | 9 | 954 | |||
BC092230.1 | - | 10 | 1028 | |||
BC141080.1 | - | 11 | 1116 | 9055692 | ||
S79780.1 | - | 10 | 1096 |