Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170779.1 | NP_740749.1 | 14 | 2071 | ||
GenBank mRNA | AB271907.1 | - | 14 | 2041 | ||
AK220259.1 | - | 2442 | ||||
BC006733.1 | - | 11 | 1660 | |||
BC017638.1 | - | 7 | 1197 | |||
BC037006.1 | - | 6 | 919 | 5116674 | ||
DQ256090.1 | - | 14 | 2071 |