Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172499.2 | NP_766087.1 | 1 | 159 | ||
GenBank mRNA | AK029874.1 | - | 1 | 160 | ||
AK169911.1 | - | 1 | 190 | |||
AK170820.1 | - | 1 | 160 | |||
BC057633.1 | - | 1 | 165 | 4035998 |