Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033245.4 | NP_001028417.1 | 17 | 2564 | ||
NM_001206390.1 | NP_001193319.1 | 17 | 2374 | |||
NM_001206391.1 | NP_001193320.1 | 17 | 2239 | |||
NM_001206392.1 | NP_001193321.1 | 16 | 2444 | |||
GenBank mRNA | AK140444.1 | - | 17 | 2428 | ||
AK149651.1 | - | 17 | 2414 | |||
AK149992.1 | - | 17 | 2376 | |||
AK152839.1 | - | 11 | 1668 | |||
AK154603.1 | - | 17 | 2536 | |||
AK155701.1 | - | 17 | 2422 | |||
AK159857.1 | - | 16 | 2290 | |||
AK159906.1 | - | 17 | 2474 | |||
AK160018.1 | - | 17 | 2474 | |||
AK160063.1 | - | 17 | 2474 | |||
AK162637.1 | - | 17 | 2507 | |||
AK163658.1 | - | 17 | 2428 | |||
AK170813.1 | - | 16 | 2294 | |||
AK171214.1 | - | 17 | 2502 | |||
AK171248.1 | - | 17 | 2414 | |||
AK171580.1 | - | 17 | 2514 | |||
BC117860.1 | - | 17 | 2459 | 40095265 | ||
BC117861.1 | - | 17 | 2324 | 40095267 |