Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AA986860 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177604.3 | NP_808272.2 | 4 | 1668 | ||
GenBank mRNA | AK028957.1 | - | 4 | 1559 | ||
AK053179.1 | - | 4 | 1670 | |||
AK085225.1 | - | 2 | 1965 | |||
AK086121.1 | - | 4 | 1543 | |||
BC099401.1 | - | 1 | 607 | 30713499 | ||
BC117765.1 | - | 1 | 881 | 40090933 | ||
BC117766.1 | - | 1 | 881 | 40090934 | ||
BC129959.1 | - | 1 | 880 | |||
BC129960.1 | - | 1 | 881 | 40090940 |