Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam193b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145382.4 | NP_663357.2 | 8 | 3068 | ||
GenBank mRNA | AK040568.1 | - | 2 | 1196 | ||
AK155237.1 | - | 8 | 3062 | |||
AK173303.1 | - | 8 | 1917 | |||
BC021381.1 | - | 2 | 658 | 5043282 | ||
BC023015.1 | - | 2 | 600 | 5364758 | ||
BC025483.1 | - | 8 | 1927 | 5150988 | ||
BC051255.1 | - | 2 | 3516 |