Gene Symbol: Wbscr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145218.3 | NP_660253.2 | 2, 3 | 1350 | ||
GenBank mRNA | AB175684.1 | - | 2, 3 | 428 | ||
AF467979.1 | - | 2, 3 | 637 | |||
AK035817.1 | - | 2, 3 | 1104 | |||
AK048758.1 | - | 2, 3 | 1095 | |||
AK051281.1 | - | 2, 3 | 1750 | |||
AK081848.1 | - | 2, 3 | 744 | |||
AK087726.1 | - | 1905 | ||||
BC052469.1 | - | 2, 3 | 746 | 6810287 | ||
BC069177.1 | - | 2, 3 | 1350 | |||
BC158110.1 | - | 2, 3 | 1377 | 9088184 |