Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190892.1 | NP_001177821.1 | 3 | 580 | ||
NM_001190898.2 | NP_001177827.1 | 4 | 789 | |||
NM_001190899.2 | NP_001177828.1 | 3 | 732 | |||
NM_001190900.1 | NP_001177829.1 | 4 | 479 | |||
NM_024411.4 | NP_077722.1 | 4 | 792 | |||
XM_011529244.1 | XP_011527546.1 | 3 | 472 | |||
XM_011529245.1 | XP_011527547.1 | 3 | 663 | |||
XM_011529246.2 | XP_011527548.1 | 3 | 650 | |||
XM_011529247.1 | XP_011527549.1 | 3 | 459 | |||
XM_011529248.1 | XP_011527550.1 | 3 | 572 | |||
XM_011529249.2 | XP_011527551.1 | 3 | 1202 | |||
XM_011529250.2 | XP_011527552.1 | 3 | 613 | |||
XM_017027878.1 | XP_016883367.1 | 3 | 462 | |||
GenBank mRNA | AK289618.1 | - | 3 | 580 | ||
AK308747.1 | - | 2 | 404 | |||
AK316076.1 | - | 4 | 429 | |||
BC026334.1 | - | 4 | 565 | 4819025 | ||
DA305883.1 | - | 4 | 479 |