Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172517.2 | NP_766105.2 | 10 | 1227 | ||
GenBank mRNA | AK049247.1 | - | 10 | 1214 | ||
AK154782.1 | - | 10 | 1228 | |||
AK170520.1 | - | 10 | 1225 | |||
AK171177.1 | - | 10 | 1232 | |||
BC030892.1 | - | 4 | 428 | |||
BC057632.1 | - | 11 | 1100 | 4918318 |