Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tet2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040400.2 | NP_001035490.2 | 4490 | |||
GenBank mRNA | AK129389.1 | - | 3 | 1677 | ||
AK151359.1 | - | 8 | 1480 | |||
AK152297.1 | - | 8 | 1480 | |||
AK153251.1 | - | 8 | 1480 | |||
AK153311.1 | - | 8 | 1480 | |||
AK153460.1 | - | 8 | 1480 | |||
BC031159.1 | - | 2 | 665 | |||
BC040785.1 | - | 2 | 665 | |||
DQ079067.1 | - | 11 | 4489 | |||
GU079949.1 | - | 9 | 4135 |