Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181328.3 | NP_851845.1 | 3 | 287 | ||
GenBank mRNA | AK047628.1 | - | 3 | 288 | ||
AK087057.1 | - | 3 | 274 | |||
AK148350.1 | - | 3 | 265 | |||
AK160906.1 | - | 3 | 236 | |||
AK181345.1 | - | 1 | 15 | |||
BC006711.1 | - | 3 | 160 | 3584570 |