Search Thermo Fisher Scientific
Gene Symbol: Zfp879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290779.1 | NP_001277708.1 | 4 | 1459 | ||
NM_173387.3 | NP_775563.1 | 5 | 1586 | |||
GenBank mRNA | AK036152.1 | - | 5 | 1587 | ||
BC118537.1 | - | 4 | 1453 | 40094138 |