Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Chtf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145412.3 | NP_663387.3 | 4 | 421 | ||
GenBank mRNA | AK030924.1 | - | 4 | 420 | ||
AK088257.1 | - | 4 | 430 | |||
AK165622.1 | - | 4 | 403 | |||
AK165993.1 | - | 2 | 247 | |||
BC023107.1 | - | 2 | 193 | 3966700 |