Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl3d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172155.1 | NP_742167.1 | 4 | 497 | ||
GenBank mRNA | AF525160.1 | - | 4 | 497 | ||
BC064735.1 | - | 4 | 502 | |||
BC119340.1 | - | 4 | 491 | 8734089 | ||
BC119344.1 | - | 4 | 491 | 8734093 |
Gene Symbol: Prl3d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205322.1 | NP_001192251.1 | 4 | 526 | ||
NM_008864.3 | NP_032890.2 | 4 | 523 | |||
GenBank mRNA | AF525162.1 | - | 497 | |||
AK082929.1 | - | 4 | 522 | |||
BC064736.1 | - | 4 | 506 | 30437709 | ||
CJ214787.1 | - | 1 | 20 | |||
M35662.1 | - | 4 | 503 |
Gene Symbol: Prl3d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172156.2 | NP_742168.1 | 497 | |||
GenBank mRNA | AF525161.1 | - | 497 | |||
BC068253.1 | - | 494 |