Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Astl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291003.1 | NP_001277932.1 | 5 | 464 | ||
NM_172539.3 | NP_766127.1 | 5 | 595 | |||
GenBank mRNA | AJ537599.2 | - | 5 | 403 | ||
AK033037.1 | - | 5 | 595 | |||
BC064729.1 | - | 5 | 456 | 30476764 | ||
DV649054.1 | - | 5 | 476 | |||
FJ187790.1 | - | 5 | 403 | |||
FJ187791.1 | - | 4 | 340 | |||
FJ187792.1 | - | 4 | 238 | |||
FJ187793.1 | - | 5 | 301 | |||
FJ187794.1 | - | 3 | 274 | |||
FJ187795.1 | - | 4 | 337 |