Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175445.4 | NP_780654.1 | 4 | 485 | ||
GenBank mRNA | AK028384.1 | - | 4 | 389 | ||
AK037601.1 | - | 4 | 341 | |||
AK172906.1 | - | 4 | 291 | |||
BC057402.1 | - | 4 | 307 | 5698333 | ||
BQ770520.1 | - | 4 | 485 |