Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178679.2 | NP_848794.1 | 3 | 1178 | ||
GenBank mRNA | AF543761.1 | - | 2 | 899 | ||
AJ516033.1 | - | 3 | 1040 | |||
AK031846.1 | - | 3 | 974 | |||
AK033892.1 | - | 3 | 940 | |||
AK044322.1 | - | 3 | 937 | |||
AK044655.1 | - | 2 | 261 | |||
AK077316.1 | - | 3 | 951 | |||
AK122387.1 | - | 3 | 1165 | |||
BC080272.1 | - | 3 | 1138 | 6837597 | ||
BC157983.1 | - | 3 | 979 | 9088140 | ||
BC157991.1 | - | 3 | 979 | 9088144 |