Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145420.2 | NP_663395.1 | 6 | 524 | ||
GenBank mRNA | AK144468.1 | - | 6 | 479 | ||
AK148364.1 | - | 6 | 512 | |||
AK214502.1 | - | 1 | 22 | |||
BC019464.1 | - | 6 | 495 | 4205941 |