Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Appl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145220.2 | NP_660255.1 | 6 | 471 | ||
GenBank mRNA | AK154525.1 | - | 6 | 495 | ||
AK160075.1 | - | 6 | 450 | |||
AK160105.1 | - | 6 | 447 | |||
AK161003.1 | - | 6 | 478 | |||
AK168403.1 | - | 6 | 432 | |||
AK168748.1 | - | 6 | 427 | |||
AK171512.1 | - | 6 | 427 | |||
AY113706.1 | - | 6 | 473 | |||
BC048906.1 | - | 6 | 409 | 6475576 |