Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zfc3h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033261.2 | NP_001028433.2 | 26 | 5207 | ||
GenBank mRNA | AK172969.1 | - | 24 | 3907 | ||
BC033596.1 | - | 4 | 537 | |||
BC061510.1 | - | 14 | 2461 | |||
BC139124.1 | - | 26 | 5064 | 8862146 |