Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285785.1 | NP_001272714.1 | 10 | 1384 | ||
NM_146011.2 | NP_666123.2 | 9 | 823 | |||
GenBank mRNA | AK047280.1 | - | 9 | 768 | ||
AK049245.1 | - | 6 | 604 | |||
AK080955.1 | - | 7 | 1739 | |||
AK083474.1 | - | 6 | 733 | |||
AK089051.1 | - | 9 | 775 | |||
AK167738.1 | - | 10 | 961 | |||
BC024535.1 | - | 9 | 818 | 4987970 | ||
BC027374.1 | - | 9 | 794 | 4947306 | ||
DQ498128.1 | - | 10 | 1232 |