Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfap36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025740.3 | NP_080016.1 | 1, 2 | 251 | ||
GenBank mRNA | AK010468.1 | - | 1, 2 | 278 | ||
AK016479.1 | - | 1, 2 | 287 | |||
AK075858.1 | - | 1, 2 | 252 | |||
AK219590.1 | - | 1, 2 | 41 | |||
BC005731.1 | - | 1, 2 | 221 | 3595965 |