Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045520.3 | NP_001038985.2 | 4 | 473 | ||
GenBank mRNA | AB093212.1 | - | 3 | 1560 | ||
AK143544.1 | - | 4 | 474 | |||
AK147875.1 | - | 4 | 458 | |||
AK168991.1 | - | 4 | 439 | |||
BC004080.1 | - | 11 |