Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045520.3 | NP_001038985.2 | 10 | 1553 | ||
GenBank mRNA | AK143544.1 | - | 10 | 1554 | ||
AK147875.1 | - | 10 | 1538 | |||
AK167454.1 | - | 2 | 165 | |||
AK168991.1 | - | 10 | 1519 | |||
AK197188.1 | - | 1 | 68 | |||
BC004080.1 | - | 7 | 1091 |