Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271356.1 | NP_001258285.1 | 12 | 1926 | ||
NM_001271357.1 | NP_001258286.1 | 12 | 1669 | |||
NM_146018.2 | NP_666130.1 | 12 | 1966 | |||
GenBank mRNA | AK039106.1 | - | 12 | 1753 | ||
AK080933.1 | - | 12 | 1751 | |||
AK136071.1 | - | 12 | 1657 | |||
AK148216.1 | - | 12 | 1720 | |||
AK154040.1 | - | 12 | 1948 | |||
AK161460.1 | - | 12 | 2009 | |||
AK172245.1 | - | 12 | 1724 | |||
AK177301.1 | - | 3 | 248 | |||
BC025820.1 | - | 12 | 1700 | 5099493 |