Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291049.1 | NP_001277978.1 | 15 | 2864 | ||
NM_173754.4 | NP_776115.2 | 15 | 2879 | |||
GenBank mRNA | AK042217.1 | - | 15 | 2865 | ||
AK047339.1 | - | 3 | 851 | |||
AK083271.1 | - | 15 | 2717 | |||
BC021474.1 | - | 14 | 2169 | 5343429 |