Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem106a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144830.3 | NP_659079.1 | 4 | 415 | ||
GenBank mRNA | AK146019.1 | - | 4 | 416 | ||
AK149650.1 | - | 3 | 659 | |||
AK150285.1 | - | 4 | 385 | |||
AK151842.1 | - | 4 | 391 | |||
AK152741.1 | - | 4 | 385 | |||
AK168437.1 | - | 4 | 358 | |||
AK169081.1 | - | 4 | 358 | |||
AK170900.1 | - | 4 | 386 | |||
AK172282.1 | - | 4 | 364 | |||
BC022145.1 | - | 3 | 463 | 5101508 |