Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC017643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252548.1 | NP_001239477.1 | 6, 7 | 752 | ||
NM_001252549.1 | NP_001239478.1 | 6, 7 | 598 | |||
NM_001252550.1 | NP_001239479.1 | 5, 6 | 698 | |||
NM_001254735.1 | NP_001241664.1 | 6, 7 | 675 | |||
NM_144832.2 | NP_659081.1 | 6, 7 | 772 | |||
GenBank mRNA | AK143743.1 | - | 4, 5 | 1789 | ||
AK145884.1 | - | 6, 7 | 738 | |||
AK158402.1 | - | 6, 7 | 709 | |||
AK159453.1 | - | 6, 7 | 730 | |||
AK168104.1 | - | 6, 7 | 730 | |||
AK170767.1 | - | 6, 7 | 720 | |||
AK185885.1 | - | 1, 2 | 59 | |||
BC017643.1 | - | 6, 7 | 728 | 4503381 | ||
BC026616.1 | - | 6, 7 | 663 | 4977003 | ||
BC027406.1 | - | 6, 7 | 557 | 4950930 |