Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L2hgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145443.2 | NP_663418.1 | 1, 2 | 224 | ||
GenBank mRNA | AK145228.1 | - | 1, 2 | 223 | ||
AK152450.1 | - | 1, 2 | 201 | |||
AK168429.1 | - | 1, 2 | 172 | |||
BC016226.1 | - | 1, 2 | 168 | 4487011 | ||
CJ087348.1 | - | 1, 2 | 226 |