Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cipc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289429.1 | NP_001276358.1 | 5 | 919 | ||
NM_001289430.1 | NP_001276359.1 | 4 | 818 | |||
NM_001289431.1 | NP_001276360.1 | 5 | 992 | |||
NM_001289432.1 | NP_001276361.1 | 5 | 866 | |||
NM_173735.3 | NP_776096.2 | 4 | 765 | |||
GenBank mRNA | AK122546.1 | - | 4 | 694 | ||
AK148415.1 | - | 3 | 695 | |||
AK154476.1 | - | 4 | 706 | |||
AK160481.1 | - | 5 | 919 | |||
AK160643.1 | - | 4 | 691 | |||
BC026384.1 | - | 4 | 697 | 4168224 |